Learn More
Trajan Scientific Rheodyne UltraLife Stator For HT/MXT715-000
Catalog No. 501037375
Encompass
Rheodyne Stator, Rheodyne, UltraLife Stator For HT/MXT715-000, UltraLife Stator is specifically designed For the Valve model HT/MXT715-000. Stator materials have been researched and selected For their physical and mechanical strengths. Stators need Replacement only if the Ports or sealing surfaces become damaged. Made in the USA.
Catalog No. 50-103-7375 Supplier Trajan Scientific Supplier No. ID.7123548
Rheodyne Stator, Rheodyne, UltraLife Stator For HT/MXT715-000, UltraLife Stator is specifically designed For the Valve model HT/MXT715-000. Stator materials have been researched and selected For their physical and mechanical strengths. Stators need Replacement only if the Ports or sealing surfaces become damaged. Made in the USA.