Learn More
Thermo Scientific™ Rheodyne™ Suction Needle Adapter
Catalog No. 03052388 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Rheodyne Injector Models 9725 and 9725i
Catalog No. For Use With (Equipment)
03-052-388 Rheodyne Injector Models 9725 and 9725i
1 options
Catalog No. 03-052-388 Supplier Thermo Scientific™ Supplier No. 9125076
Add to Cart
Add to Cart
Enable suction loading of Thermo Scientific™ Rheodyne™ 9725 and 9725i Sample Injectors for HPLC with this adapter.
Use the Thermo Scientific™ Rheodyne™ Suction Needle Adapter for suction loading of samples into the Rheodyne 9725 and 9725i Sample Injectors for high-performance liquid chromatography (HPLC),
TRUSTED_SUSTAINABILITY
Specifications
Type Suction Needle Adapter
For Use With (Application) Liquid Chromatography
Description Suction Needle Adapter
For Use With (Equipment) Rheodyne Injector Models 9725 and 9725i
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.