Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Rheoscopic Concentrate
SCI_ED

Catalog No. S37741 Shop All Aldon Products
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S37741 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

WARNING: This product can expose you to chemicals including Ethylene Oxide, which is known to the State of California to cause cancer and reproductive harm.

Catalog No. S37741 Supplier Innovating Science™ Supplier No. IS5061
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Ethylene Oxide, which is known to the State of California to cause cancer and reproductive harm.

Pearly-white, water based solution. Innovating Science Rheoscopic solution can be used to demonstrate concepts that are usually difficult to see such as: ocean currents, turbulence, and convection.

  • Add food coloring to color the solution which will make it even easier and more exciting to simulate oceanic and atmospheric patterns
  • 120 mL concentrate makes 7.5L

Specifications

Grade 3+
Product Type Rheoscopic Fluid
Includes 120 mL concentrate
Weight (English) 1 lb.
For Use With (Application) To demonstrate concepts that are usually difficult to see such as: ocean currents, turbulence, and convection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.