Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Rheoscopic Solution
SCI_ED

Catalog No. S06634 Shop All Aldon Products
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06634 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

WARNING: This product can expose you to chemicals including Ethylene Oxide, which is known to the State of California to cause cancer and reproductive harm.

Catalog No. S06634 Supplier Innovating Science™ Supplier No. IS5060
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Ethylene Oxide, which is known to the State of California to cause cancer and reproductive harm.

Pearly white, water-based solution to demonstrate ocean currents, turbulence and convection, which are usually difficult to see.

  • Addition of food colors to the solution to see simulated oceanic and atmospheric patterns.
  • Water-based suspension.
  • Nontoxic, non-flammable.
  • Unlimited shelf life.

Specifications

Grade 3+
Product Type Rheoscopic Fluid
Includes 1 L Rheoscopic Fluid
Weight (English) 2.3 lb.
For Use With (Application) To demonstrate ocean currents, turbulence and convection, which are usually difficult to see
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.