Learn More
Eurofins Discoverx Recombinant Human Heregulin β1, 10 µg. (92-1031)
Supplier:  Eurofins Discoverx 921031
Encompass
Heregulin β-1 is a signaling protein for the ErbB2/ErbB4 and ErbB2/ErbB3 receptor heterodimers. Plays an important role in the heart structure and function by inducing receptor phosphorylation and cardiomyocyte differentiation. Proteolytic processing of the extracellular domain of the transmembrane NRG-1 isoforms release soluble growth factor versions. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9577