Learn More
Sino Biological Recombinant Rhesus ACE2 Protein (hFc Tag)
Supplier:  Sino Biological 90211C02H100
Encompass_Preferred
A DNA sequence encoding the rhesus ACE2 (ACI04564.1) (Met1-Val739) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2706