Learn More
Sino Biological Rhesus ACE2 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 90211C02H20
Encompass_Preferred
A DNA sequence encoding the rhesus ACE2 (ACI04564.1) (Met1-Val739) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1666