Learn More
Sino Biological Rhesus AGRP Protein 200ug (Sino Biological)
Supplier:  Sino Biological 90196C04H200
Encompass_Preferred
A DNA sequence encoding the rhesus AGRP (XP_001091740.1) (Ser83-Thr132) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-237-3259