Learn More
Sino Biological Rhesus AMY2A Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90012C02H100
Encompass_Preferred
A DNA sequence encoding the rhesus AMY2A (H9EX43) (Met1-Leu511) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2589