Learn More
Bon Opus Biosciences Recombinant Rhesus CCL1 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP108450UG
Encompass_Preferred
Recombinant Rhesus CCL1 (C-His) is produced by Human Cells. The target gene encoding Lys24-Lys96 is expressed with a C-His tag. Accession# Q8HYQ4. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1084
Catalog No. 50-217-2682