Learn More
Sino Biological Rhesus CCL21 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 90138MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus CCL21 / 6Ckine (Catalog#90138-CNAB; NP_001028027.1; Met1-Pro131). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2695