Learn More
Sino Biological Rhesus CD160 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90224C08H100
Encompass_Preferred
A DNA sequence encoding the rhesus CD160 (XP_001089019.1) (Met1-Leu158) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1851