Learn More
Sino Biological Rhesus CD160 Protein 100ug (Sino Biological)
Catalog No. 502374198
Encompass_Preferred
A DNA sequence encoding the rhesus CD160 (XP_001089019.1) (Met1-Leu158) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-237-4198 Supplier Sino Biological Supplier No. SEKA1094715
A DNA sequence encoding the rhesus CD160 (XP_001089019.1) (Met1-Leu158) was expressed with six amino acids (LEVLFQ) at the C-terminus.