Learn More
Sino Biological Rhesus CD1A & B2M Protein 100ug (Sino Biological)
Supplier:  Sino Biological 13124H01H50
Encompass_Preferred
A DNA sequence encoding the rhesus CD1A(NP_001139290.1) (Met1-Ser299) of was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the rhesus B2M (NP_001040602.1) (Met1-Met119) constructed the plasmid 2. The two plasmids were co-expressed and the rhesus CD1C/B2M heterodimer was purified.
Catalog No. 50-237-4039