Learn More
Sino Biological Rhesus CD209 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90319C01H50
Encompass_Preferred
A DNA sequence encoding the rhesus CD209 (AAK74185.1) (Lys62-Glu381) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-2116