Learn More
Sino Biological Cynomolgus / Rhesus TNFR2 / CD120b / TNFRSF1B Protein (His Tag) 1mg
Supplier:  Sino Biological 90102C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus TNFRSF1B (XP_005544817.1/NP_001253134.1) (Met1-Asp257) was expressed, fused with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus TNFRSF1B sequences are identical.
Catalog No. 50-222-3810