Learn More
Sino Biological Rhesus CD22 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90246C02H50
Encompass_Preferred
A DNA sequence encoding the rhesus CD22 (EHH29920.1) (Met1-Arg686) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2270