Learn More
Sino Biological Recombinant Rhesus CD3 epsilon/CD3e Protein (His Tag)
Supplier:  Sino Biological 90047C08H500
Encompass_Preferred
A DNA sequence encoding the rhesus CD3E (XP_001097204.1) CD3E extracellular domain (Met 1-Asp 117) was fused with a polyhistidine tag at the C-terminus.
Catalog No. NC2874181