Learn More
Sino Biological Rhesus CD32A/FCGR2A Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785782
Encompass_Preferred
The Rhesus CD32a / Fc gamma RIIA / FCGR2A ELISA Pair Set is for the quantitative determination of Rhesus CD32a / Fc gamma RIIA / FCGR2A.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD32a / Fc gamma RIIA / FCGR2A coated on a 96-well plate. Standards and samples are added to the wells, and any CD32a / Fc gamma RIIA / FCGR2A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-CD32a / Fc gamma RIIA / FCGR2A monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD32a / Fc gamma RIIA / FCGR2A present in the sample. To end the enzyme reaction, the stop solution is added and absorban
Catalog No. 50-007-85782 Supplier Sino Biological Supplier No. SEK9001615
The Rhesus CD32a / Fc gamma RIIA / FCGR2A ELISA Pair Set is for the quantitative determination of Rhesus CD32a / Fc gamma RIIA / FCGR2A.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD32a / Fc gamma RIIA / FCGR2A coated on a 96-well plate. Standards and samples are added to the wells, and any CD32a / Fc gamma RIIA / FCGR2A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-CD32a / Fc gamma RIIA / FCGR2A monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD32a / Fc gamma RIIA / FCGR2A present in the sample. To end the enzyme reaction, the stop solution is added and absorban