Learn More
Sino Biological Anti-Siglec-3/CD33 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90303RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus / Rhesus CD33 / Siglec-3 (Catalog#90303-C08H; EHH59833.1 / EHH30318.1; Met1-Gly248). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0484