Learn More
Sino Biological Recombinant Rhesus CD3D & CD3E Heterodimer Protein (ECD, Fc-Flag & Fc-His Tag), HPLC-verified
Supplier:  Sino Biological CT032C0323H4500
Encompass_Preferred
A DNA sequence encoding the Rhesus CD3D (F6WI60-1)(Met1-Ala105) was fused with the C-terminal flag-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the Rhesus CD3E (XP_014971302.1) (Met1-Asp117) with the C-terminal his-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the CD3D&CD3E heterodimer was purified.
Catalog No. 50-224-6573