Learn More
Sino Biological Anti-CD4 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 90274MM53F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus CD4 ( Catalog#90274-C08H; EHH20435.1; Met1-Trp390) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8382