Learn More
Sino Biological Recombinant Rhesus CD4 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 90274C02H100
Encompass_Preferred
A DNA sequence encoding the rhesus CD4 (EHH20435.1) (Met1-Trp390) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2688