Learn More
Sino Biological Rhesus CD40LG Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90096C01H100
Encompass_Preferred
A DNA sequence encoding the rhesus CD40LG (EHH31125.1) (Met113-Leu261) was expressed, with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-3390