Learn More
Sino Biological Recombinant Anti-IL-8/CXCL8 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 90280R009100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rhesus IL-8 / Interleukin-8 / CXCL8 ( Catalog#90280-CNAE; NP_001028137.1; Ala23-Pro101).
Catalog No. 50-240-8636