Learn More
Sino Biological Anti-IL-8/CXCL8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90280RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus IL-8 / CXCL8 (Catalog#90280-CNAE; NP_001028137.1; Ala23-Pro101). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2196