Learn More
Sino Biological Rhesus EFNA5 Protein 200ug (Sino Biological)
Supplier:  Sino Biological 90044C08H200
Encompass_Preferred
A DNA sequence encoding the rhesus EFNA5 (NP_001252657.1) (Met1-Asn203) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3706