Learn More
Sino Biological Rhesus EGFR PAb 100ul (Sino Biological)
Supplier:  Sino Biological 90317RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus EGFR / HER1 / ErbB1 (Catalog#90317-K08H; XP_001107305.1; Met1-Ser645). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-1561