Learn More
Sino Biological Rhesus EphA4 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90039C02H100
Encompass_Preferred
A DNA sequence encoding the rhesus EPHA4 (NP_001247799.1) (Met1-Thr547) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2300