Learn More
Sino Biological Rhesus EphB1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90040C08H100
Encompass_Preferred
A DNA sequence encoding the rhesus EPHB1 (XP_001115263.1) (Met1-Pro540) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3564