Learn More
Sino Biological Anti-Her2/ERBB2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90020RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus HER2 / ErbB2 (rh ERBB2; Catalog#90020-K08H; XP_001090430.1; Met1-Thr652). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2579