Learn More
Sino Biological Anti-Her2/ERBB2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90020RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus HER2 / ErbB2 (Catalog#90020-K08H; XP_001090430.1; Met 1-Thr 652). HER2 / ErbB2 specific IgG was purified by Rhesus HER2 / ErbB2 affinity chromatography.
Catalog No. 50-240-8939