Learn More
Sino Biological Anti-HER3/ERBB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90043T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus HER3 / ErbB3 (rh HER3 / ErbB3; Catalog#90043-K08H; XP_005571238.1 / XP_001113953.2; Met1-Thr643). HER3 / ErbB3 specific IgG was purified by Rhesus HER3 / ErbB3 affinity chromatography.
Catalog No. 50-240-9898