Learn More
Sino Biological Rhesus FCGR2A Protein 20ug (Sino Biological)
Supplier:  Sino Biological 90016K49HB20
Encompass_Preferred
A DNA sequence encoding the Rhesus FCGR2A (NP_001244229.1) (Met1-Ile217) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-3685