Learn More
Sino Biological Recombinant Rhesus CD16/FCGR3/Fc gamma RIII Protein (His & AVI Tag)
Supplier:  Sino Biological 90013C27H1
Encompass_Preferred
A DNA sequence encoding the rhesus CD16 (NP_001258584.1) extracellular domain (Met 1- Gln 208) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus.
Catalog No. 50-255-2746