Learn More
Sino Biological Rhesus FCN1 Protein 200ug (Sino Biological)
Supplier:  Sino Biological 90154C02H200
Encompass_Preferred
A DNA sequence encoding the rhesus FCN1 (XP_001098002.1) (Met1-Ala326) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3033