Learn More
Sino Biological Recombinant Anti-FGFR4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 90134R148100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rhesus FGFR4 / CD334 ( Catalog#90134-C08H; XP_001087243.1; Met1-Asp369).
Catalog No. 50-240-8889