Learn More
Sino Biological Anti-FGFR4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90134RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus FGFR4 / CD334 ( Catalog#90134-C08H; XP_001087243.1; Met1-Asp369). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8890