Learn More
Sino Biological Rhesus FGFR4 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90134C08H100
Encompass_Preferred
A DNA sequence encoding the rhesus FGFR4 (XP_001087243.1) (Met1-Asp369) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1522