Learn More
Sino Biological Recombinant Cynomolgus PD-1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90311C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus PDCD1 (NP_001271065.1) (Met1-Gln167) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-222-3841