Learn More
Sino Biological Anti-Interferon alpha/beta receptor 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90113T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus IFNAR1 ( Catalog#90113-C08H; NP_001253442.1; Met1-Lys436). IFNAR1 specific IgG was purified by Rhesus IFNAR1 affinity chromatography.
Catalog No. 50-255-2717