Learn More
Sino Biological Anti-IGFBP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90083T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus IGFBP-2 / IGFBP2 (Catalog#90083-C08H; XP_001087071.1; Met1-Gln324). IGFBP-2 / IGFBP2 specific IgG was purified by Rhesus IGFBP-2 / IGFBP2 affinity chromatography.
Catalog No. 50-255-2724