Learn More
Sino Biological Recombinant Rhesus IL-1 beta Protein
Supplier:  Sino Biological 90010CNAE20
Encompass_Preferred
A DNA sequence encoding the mature form of rhesus IL1B (NP_001036221.1) (Ala117-Ser269) was expressed and purified, with an initial Met.
Catalog No. 50-228-2006