Learn More
Sino Biological Recombinant Rhesus IL-12 Protein (His Tag) 50
Supplier:  Sino Biological CT045C08H50
Encompass_Preferred
A DNA sequence encoding the rhesus IL12A (NP_001038199.1) (Met1-Ser219) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the rhesus IL12B (NP_001038190.1) (Met1-Ser328) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the rhesus IL12A & the rhesus IL12B heterodimer was purified.
Catalog No. 50-224-6678