Learn More
Sino Biological Rhesus IL12A Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90290C02H50
Encompass_Preferred
A DNA sequence encoding the rhesus IL12A (NP_001038199.1) (Met1-Ser219) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1996