Learn More
Sino Biological Rhesus IL17RA Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90127C08H100
Encompass_Preferred
A DNA sequence encoding the rhesus IL17RA (XP_001102483.2) (Met1-Trp320) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3654