Learn More
Sino Biological Recombinant Rhesus IL-18R beta / IL-18RAP Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 90122C08H1
Encompass_Preferred
A DNA sequence encoding the rhesus IL18RAP (XP_001108108.1) (Met1-Arg356) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6910