Learn More
Sino Biological Recombinant Rhesus CD25/IL-2R alpha Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 90265C27HB100
Encompass_Preferred
A DNA sequence encoding the rhesus IL2RA (NP_001028089.1) (Met1-Arg213) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2693