Learn More
Sino Biological Rhesus IL6ST PAb 100ul (Sino Biological)
Supplier:  Sino Biological 90275T10100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus IL6ST / gp130 / CD130 ( Catalog#90275-C08H; NP_001252920.1; Met1-Ile681). IL6ST / gp130 / CD130 specific IgG was purified by Rhesus IL6ST / gp130 / CD130 affinity chromatography.
Catalog No. 50-237-3147