Learn More
Sino Biological Recombinant Rhesus gp130/IL6ST Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 90275C02H1
Encompass_Preferred
A DNA sequence encoding the rhesus IL6ST (NP_001252920.1) (Met1-Ile618) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2687